Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ arium™ Pro ASTM TYPE 1 - Ultrapure Water System: Wall Mounted

Produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis with the Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System.

Manufacturer:  Sartorius™ H2OPROUVB

 View more versions of this product

Catalog No. 14-556-216


Add to cart

Description

Description

The arium pro series consist of flexible systems with device configurations that can be specially custom-tailored to applications and feature an outstanding cost-benefit ratio. All systems meet and exceed ASTM Type 1 water quality and ensure the best, reproducible results. Per hour, an arium pro can deliver up to 120 l of ultrapure water with a conductivity of 0.055µS/cm (18.2 megaohm/cm at 25°C) and consistently high quality. When a Sartopore° 2 150 final filter is used, arium pro will deliver ultrapure water that is practically free of microorganisms.

  • Mounts on wall
  • Multiple configurations available based on application needs
  • Optimized components and cartridge materials with topdown-flow technology
  • Optional, integrated UV lamp (185/254nm) for eliminating bacteria and organics; controlled by reliable TOC monitoring
  • Display with touch-activated functions and intuitive menu navigation
  • Integrated real time monitoring and data display: feed conductivity, total water consumption, pressure regulator
  • PIN-protected access to basic settings /service
  • Convenient SD card slot

  • System can be chosen based on daily work
  • Guarantees a long service life and optimal water purification
  • Reliable water quality ensures reproducible results
  • Easy operation; no user manual needed
  • Optimal control of constant water flow and consistent water quality without extra costs
  • Prevents user operating errors
  • Supports reliable recording of important data

Specifications

Specifications

Type 1
120
Less than2 Product water, Less than50 feed water
19.37 in.
13.7 in.
100/240V
35.6 to 95°F
17.75 in.
Less than1
ASTM
100/240V 50/60Hz
2 to 30°C
Wall mounting kit, dispenser unit with 2.8m extended cable, tubing and wall bracket
35.6 to 86°F
2
0.055
59.5 to 63.9 lb.
Arium pro System with UV Lamp
18.2
≤2
Less than1CFU/1000mL
49.2cm
35cm
50/60Hz
2 to 35°C
45.1cm
5 to 45°C
Chromatographic
80%
4 to 10
Less than1/mL
Wall
130
Adsorption by spherical activated Carbon, deionization, optional final particle and sterile filtration
27 to 29kg
Documents
Promotions

Promotions

  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit