Sartorius™ arium™ Pro ASTM TYPE 1 - Ultrapure Water System: Wall Mounted

Produce ASTM type 1 water for critical analysis like HPLC, PCR, antibody production, cell culture media preparation or electrophoresis with the Sartorius arium™ Pro ASTM TYPE 1 - Ultrapure Water System.

Manufacturer: Sartorius™ H2OPRODIB

 View more versions of this product

Catalog No. 14-555-486

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Product Type Arium pro System-DI
    Product Water Type Type 1
    Height (Metric) 49.2cm
    Resistivity 18.2
    Hertz 50/60Hz
    Temperature Range (English) 35.6 to 95°F
    Temperature Range (Metric) 2 to 35°C
    Depth (English) 17.75 in.
    Depth (Metric) 45.1cm
    Daily Output 120
    Turbidity Less than1
    Storage Requirements 5 to 45°C
    Certifications/Compliance ASTM
    For Use With (Application) General
    Flow Rate ≤2
    Electrical Requirements 100/240V 50/60Hz
    Humidity Range 80%
    Feedwater Temperature (Metric) 2 to 30°C
    pH Range 4 to 10
    Particle Size Less than1/mL
    Total Organic Carbon Less than5 Product water, Less than50 feed water
    Feedwater Temperature (English) 35.6 to 86°F
    Mounting Wall
    Liquid Flow Rate 2
    Power Supply Details 130
    Range (Conductivity) 0.055
    Purification Method Adsorption by spherical activated Carbon, deionization, optional final particle and sterile filtration
    Weight (English) 59.5 to 63.9 lb.
    Weight (Metric) 27 to 29kg
    Bacterial Content Less than1CFU/1000mL
    Height (English) 19.37 in.
    Width (English) 13.7 in.
    Width (Metric) 35cm
    Voltage 100/240V

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit