Learn More
Sartorius™ Sartorius™ Balance Dot-Matrix Printers
Specifications
Type Dot Matrix Printer
View More Specs
Includes: Supplied AC adapter/charger also permits AC operation at 115V 50/60Hz. Comes with connection cable, one roll of paper, and one purple ink cartridge.
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01-913-377
View Documents
Sartorius™
901041.1
DI, TL, TR, TC Series This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Dot-Matrix Printer. For Sartorius balances. Ink ribbon. Prints up to 1 3/4 lines per second; 40 characters per line. Standard RS-232-C interface. Adjustable baud rate between 300 and 19,200bps. Features full dot addressable graphics, alphanumeric characters (including an international character set) for printing weighing units or statistical data, and an integral self-test and diagnostic mode.
Order Info
Unit measures 7 3/4L x 4 1/8W x 3 1/2 in.H (19.7 x 10.5 x 8.9cm). Net weight 1 3/8 lb. (0.6kg).
Specifications
Specifications
Dot Matrix Printer