Sartorius™ Interface Cables for SE and ME Ultramicro and Microbalances Available on GSA/VA Contract for Federal Government customers only.
For use with SE and ME Ultramicro and Microbalances.
Manufacturer: Sartorius™ YDO01ME
 View more versions of this product
Catalog No. 14-557-511
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SE and ME ultramicro and microbalance
    Product Type Interface Cable
    Connection Type Interface 9 pin
    Type Interface Cable
    • Data interface
    • PC Compatible
    • Interface 9 pin
    • Incl. 5-contact DIN connector for bar code scanner or keyboard
  • Description & Specifications
Related Products
Product Content Feedback