Sartorius™ Interface Cables for SE and ME Ultramicro and Microbalances Available on GSA/VA Contract for Federal Government customers only.
For use with SE and ME Ultramicro and Microbalances.
Manufacturer: Sartorius™ YDO02ME
 View more versions of this product
Catalog No. 14-557-512
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SE and ME ultramicro and microbalance, barcode scanner or PC keyboard
    Product Type Interface Cable
    Connection Type RS485 data interface, 12 pin
    Type Interface Cable
    • RS-485
    • 12-pin round
    • With connector for bar code scanner or keyboard
  • Description & Specifications
Related Products
Product Content Feedback