Learn More
Sartorius™ Balance Printers
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
Manufacturer:  Sartorius™ YDP10BT0CE
 View more versions of this product
Catalog No. 14-557-542
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Built-in real-time clock
  • Built-in statistics program
  • Counter for consecutive numbering of transactions can be activated
  • High contrast LCD shows date or time as well as current transaction number
  • Integrated rechargeable batteries with battery level indicator
  • Ultrafast print head
  • Runs on the balance's power; no separate power supply necessary
  • Easy to change paper roll and ink ribbon cassette
Specifications
Specifications
Data Printer
230 x 158 x 80mm
158mm
Bluetooth™ radio interface, Paper roll, fou
Cubis™ balance, bluetooth wireless radio int
230mm
80mm
0.77kg
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021