Learn More
Sartorius™ Balance and Scale Weighing Pans Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 69V00004
 View more versions of this product
Catalog No. 14-557-597
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Stainless Steel
965mm
For models AY123, -303, -212
Weighing Pan
3.8 in.
Weighing samples
Square