Sartorius™ Balance Wedge and Wedge Pro Data Transfer Software Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Sartorius™ YSW06
 View more versions of this product
Catalog No. 14-557-515
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance,CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance
    Product Type Wedge Pro
    For Use With (Application) Completely collects balance data and transfers it to any DOS/Windows application program
    Includes 5 ft. RS232 cable
    Type Wedge Pro
    • For Windows
    • With Ethernet connectivity and 5 ft. cable
  • Description & Specifications
Related Products
Product Content Feedback