Sartorius™ Balance Wedge and Wedge Pro Data Transfer Software
Manufacturer: Sartorius™ YSW05
 View more versions of this product
Catalog No. 14-557-514
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Equipment) CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance,CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance
    Includes 5 ft. RS232 cable
    Product Type Wedge
    For Use With (Application) Completely collects balance data and transfers it to any DOS/Windows application program
    Type Wedge
  • Description & Specifications
Related Products
Product Content Feedback