Sartorius™ Biohit™ Adapter Sets for Midi Plus™ Electronic Pipetting Controller
Adapter sets for Midi Plus Electronic Pipetting Controller
Manufacturer: Sartorius™ 711015
 View more versions of this product
Catalog No. 14-559-564
  Check Availability
Add to Cart
    Specifications
    Autoclavable Yes
    For Use With (Equipment) Midi Plus™ electronic pipetteting controller with Biohit™ design
    Includes Nose cone and the Silicon adapter
    Product Type Adapter Set for 5mL tips
  • Description & Specifications
Related Products
Product Content Feedback