Learn More
Sartorius™ Biohit™ Dispenser Tips
Convenient and ergonomic approaches for repetitive dispensing
Manufacturer:  Sartorius™ 792021
 View more versions of this product
Catalog No. 14-559-532
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For use with eLINE™ Pro, Lite Dispensers, Biohit Mechanical Stepper, (Ripette (Ritter), HandyStep (Brand), Minilab 100/101 (HTL), EasyStep)F
  • Function according to positive displacement principle
  • Contamination-free dispensing
  • No aerosols formed
  • Made of virgin polypropylene (tip) and polyethylene (plunger)
  • Nine different tip sizes from 0.1 to 50mL
  • DNA, RNase, ATP and endotoxin-free
  • Pre-sterilized tips are individually wrapped
Certifications
CE marked
Specifications
Specifications
Sartorius™ Biohit™ eLINE™, Sartorius™ Biohit™ eLINE™ Lite, Sartorius™ Biohit™ eLINE™ Pro, Universal Fit Repeater Pipettor
Non-sterile
PE (Polyethylene)/PP (Polypropylene)
Sartorius™ Biohit™ eLINE™
2.5mL
Yes
CE
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021