Sartorius™ Biohit™ Dispenser Tips
Convenient and ergonomic approaches for repetitive dispensing
Manufacturer: Sartorius™ 792023
 View all options for this product
Catalog No. 14-559-536
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Product Type Dispenser Tip
    Volume (Metric) 10mL
    Sterility Nonsterile
    Autoclavable Yes
    Material Polyethylene/Polypropylene
    Quantity 100
    Certifications/Compliance CE
    For Use With (Equipment) eLINE™ Pro and Lite dispensers, mechanical steppers
    For use with eLINE™ Pro, Lite Dispensers, Biohit Mechanical Stepper, (Ripette (Ritter), HandyStep (Brand), Minilab 100/101 (HTL), EasyStep)F
    • Function according to positive displacement principle
    • Contamination-free dispensing
    • No aerosols formed
    • Made of virgin polypropylene (tip) and polyethylene (plunger)
    • Nine different tip sizes from 0.1 to 50mL
    • DNA, RNase, ATP and endotoxin-free
    • Pre-sterilized tips are individually wrapped
  • Description & Specifications
Related Products
Product Content Feedback