Learn More
Sartorius™ Biohit™ eLINE™ Multichannel Electronic Pipet
Handles several types of pipetting tasks from regular pipetting to diluting with mixing and automatic repetitive dispensing. Sartorius™ Biohit™ eLINE™ Multichannel Electronic Pipets with fully electronic operations guarantee user-independent results and high ergonomics to prevent Repetitive Strain Injury.
Specifications
Certifications/Compliance CE, IVD
Finger Style Ergonomic
Quantity Each
Warranty 2 Years
View More Specs
Products 1
Catalog Number Mfr. No. Accuracy Color Number of Channels Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Accuracy Color Number of Channels Volume (Metric) Price Quantity & Availability  
14-559-391
View Documents
Sartorius™
730491
0.5%, 1%, 2.5% Purple 12 50 to 1200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Electronic pipet made to deliver years of real performance and total user convenience features a comprehensive range of pipetting modes and is twice as fast as a mechanical pipete.
  • Unique DC motor concept provides outstanding accuracy and precision
  • Fully electronic operations guarantee user-independent results and prevent RSI
  • Effortless, electronic tip ejection
  • Ensures contamination-free pipetting
  • Available with several charging options (charging stand, carousel and direct charging)
  • Built-in error control for more reliable results
  • Ergonomic design suits both left- and right-handed operation
  • Optiload mechanism allows perfect tip sealing
  • Easy programming; six memory places for favorite programs
  • Single point calibration for special liquids
  • Autoclavable dispensing heads (1200µL, 8/12 channel models)
  • Safe-Cone Filters help to prevent contamination and damage of pipet
  • One eLINE covers the volume range of two mechanical pipets
  • Comprehensive range of pipetting programs
Certifications
CE marked
Warranty and Services
2 years
Specifications
Specifications
CE, IVD
Ergonomic
Each
2 Years