Sartorius™ Biohit™ eLINE™ Multichannel Electronic Pipet
Handles several types of pipetting tasks from regular pipetting to diluting with mixing and automatic repetitive dispensing. Sartorius™ Biohit™ eLINE™ Multichannel Electronic Pipets with fully electronic operations guarantee user-independent results and high ergonomics to prevent Repetitive Strain Injury.
Manufacturer: Sartorius™ 730420
 View all options for this product
Catalog No. 14-559-384
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Accuracy 0.9%, 1.5%, 4%
    Certifications/Compliance CE, IVD
    Color Gray
    Color Code Gray
    Number of Channels 12
    Volume (Metric) 0.2 to 10μL
    Finger Style Ergonomic
    Increments 0.05μL
    Packaging Quantity Ea.
    Precision 3.00 to 0.50
    Warranty 2 Years
    Electronic pipet made to deliver years of real performance and total user convenience features a comprehensive range of pipetting modes and is twice as fast as a mechanical pipete.
    • Unique DC motor concept provides outstanding accuracy and precision
    • Fully electronic operations guarantee user-independent results and prevent RSI
    • Effortless, electronic tip ejection
    • Ensures contamination-free pipetting
    • Available with several charging options (charging stand, carousel and direct charging)
    • Built-in error control for more reliable results
    • Ergonomic design suits both left- and right-handed operation
    • Optiload mechanism allows perfect tip sealing
    • Easy programming; six memory places for favorite programs
    • Single point calibration for special liquids
    • Autoclavable dispensing heads (1200µL, 8/12 channel models)
    • Safe-Cone Filters help to prevent contamination and damage of pipet
    • One eLINE covers the volume range of two mechanical pipets
    • Comprehensive range of pipetting programs
  • Description & Specifications
Related Products
Product Content Feedback