Learn More
Sartorius™ Biohit™ eLINE™ Single-Channel Electronic Pipet
Pipet twice as fast as with a mechanical pipet. Sartorius™ Biohit™ eLINE™ Single-Channel Electronic Pipet features multiple modes to streamline lab work and enable fast dispensing. Delivers years of real performance and total convenience. Available in a range of volumes, from 0.1µ to 5000µL.
Specifications
Battery Type NiMH
Calibration Points Single
Certifications/Compliance CE
Model Left and right hand
Display LCD
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Increments Includes Precision Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Increments Includes Precision Price Quantity & Availability  
14-559-367
View Documents
Sartorius™
730021
0.2 to 10 μL 0.05μL AC adapter 10.00 to 0.40 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-375
View Documents
Sartorius™
730101
100 to 5000 μL 10μL AC adapter 0.40 to 0.15 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Unbeatable performance and accuracy
  • Unique DC motor concept provides unbeatable accuracy and precision
  • Enables liquid dispensing significantly faster than with a mechanical pipette.
  • Electronic operations guarantee user-independent results and prevents RSI
  • Built-in error control provides more reliable results
  • Single point calibration for special liquids
  • Nine speeds for both aspiration and dispensing
  • Can be used for several liquid handling tasks, from pipetting to diluting and automatic dispensing
  • Autoclavable dispensing heads
Convenient and user-friendly
  • Effortless, electronic tip ejection with ergonomic design
  • Ergonomic design that suits both left- and right-handed
  • Easy programming, six memory places for favorite programs
  • Volume range color coding for easy pipet and tip selection
  • Different charging options (charging stand, charging carousel and direct charging)
  • Available with or without AC adapter (EU, UK, US/JPN and CHN plugs)
  • Clear LCD-display; easy-to-use keypad with electronic start button
  • Easy to clean and maintain
  • Low battery indicator
Super Pipeting Feature ( Available in 0.1-5µL model)
  • Enables dispensing of the smallest volumes from the air, emptying tip completely
  • Increased liquid dispensing force empties the tip, with no drops left hanging
  • Prevent droplets from clinging onto the electrically charged outer surface of microplate well
  • Increases accuracy of the dispensed volume
  • Less tip ejections; savings in tip costs
  • Particularly useful for PCR and microplate pipetting
Certifications
CE marked
Warranty and Services
Two years
† Where a range is given, the first value applies to the smallest volume in the stated range; the second, to the largest volume. (Exception: for the 0.1 to 5µL pipetter, the first value is for a dispensed volume of 0.5µL.)
Specifications
Specifications
NiMH
CE
LCD
2 Years
Single
Left and right hand
15°C to 40°C