Sartorius™ Biohit™ mLINE™ Pipette Multipacks
Replenish or set up a new pipetting station. Sartorius™ Biohit™ mLINE™ Pipette Multipacks conveniently combine a set of mLINE mechanical pipettes with matching tip trays and useful accessories. Available in collections of 3, 4 and 5 pipettes with matching tips
Manufacturer: Sartorius™ LH725661
 View more versions of this product
Catalog No. 14-557-700
  Check Availability
Add to Cart
    Specifications
    Includes Pipets: 10μL, 100μL and 1,000μL, single channel
    Optifit Tip tray boxes: 10μl, 200μL and 1,000μL
    Linear stand and accessories
    ,Pipets: .05 to 10uL, 10 to 100uL, 100 to 1000uL (one each), Sartorius Optifit Tip tray box: 0.2 to 10ul, 0.5 to 200ul, 10 to 1000ul (one each), linear stand
    Product Type Pipette 3-pack (10/100/1000)
    Autoclavable Yes
    Description Combines mLINE pipets and Optifit Tips with for safe, precise pipetting
    For Use With (Application) Repetitive manual pipetting
    Packaging Quantity 3 pipettes with 3 boxes of Optifit Trays
    Number of Channels 1 per pipette
    Type 3-Pack 10
    Multipacks to match every need
    • Affordable options to get a set of quality pipettes with useful accessories
    • Ideal for renewing an existing pipette collection or setting up a new pipetting station
    • Includes a linear pipette rack, calibration tool, tube opener and color-coding caps
    • Collections include up to five different volumes of single channel pipettes, from 10µL to 5mL; the 3+1 pack includes a 350µL eight-channel pipette
    mLINE&trade mechanical pipettes
    • Offer excellent ergonomics, performance, precision and safety in manual pipetting
    • Requires exceptionally light pipetting and tip ejection forces
    • Specifically designed to prevent Repetitive Strain Injury (RSI) in repetitive pipetting
    • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
    • Increased safety with volume lock, preventing accidental volume changes while pipetting
    Optifit Tips single trays
    • Designed for the best possible fitting and sealing for the highest possible accuracy and precision
    • Tips are RNase, DNase and endotoxin free and compliant with ISO 9001 and ISO 13485
    • Environment-friendly materials and fully autoclavable to reduce waste
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback