Sartorius™ Biohit™ Optifit 10 to 1000uL Tips, Bulk in a Box

The perfect match for your pipetter

Manufacturer: Sartorius™ 791005

 View more versions of this product

Catalog No. 14-559-482

  Check Availability
Add to Cart

    Specifications

    Product Type Optifit Tip
    Material Polypropylene
    Quantity Bulk in box
    Length (English) 2.81 in.
    Length (Metric) 7.15cm
    For Use With (Equipment) Optiload feature in Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipetters
    Sterility Nonsterile
    Autoclavable Yes
    Packaging Bulk Box
    Certifications/Compliance DNase, RNase and endotoxin-free, CE, ISO 9001, ISO 13485
    Volume (Metric) 10 to 1000μL
    Color Blue
    Color Code Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit