Sartorius™ Biohit™ Pipet Charging Stands
Accessory for in vitro diagnostic product
Manufacturer: Sartorius™ 511601
 View more versions of this product
Catalog No. 14-559-397
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Number of Channels 4.0
    For Use With (Equipment) Proline™ electronic pipette
    Includes Universal AC adapter
    • Bench top charging units
    • Maintenance free: Regular cleaning ensures trouble-free operation
    • AC Adaptor: Output voltage 9 VDC
    The Biohit Proline Charging Stand/Carousel is intended to be used only with Biohit Proline Electronic pipetors. To guarantee optimal charging, do not use any other than the original Biohit AC-adaptor.
  • Description & Specifications
Related Products
Product Content Feedback