Learn More
Sartorius™ Biohit™ Pipet Stands and Accessories
Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius™ Biohit™ Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.
Manufacturer:  Sartorius™ 726001
 View more versions of this product
Catalog No. 14-559-422
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Pipette Stands
  • Stores pipettes in an upright position while not in use to avoid contamination from the desktop
    • Linear Stand is designed for all Biohit mechanical devices and stores them neatly in a line for instant access
    • Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
    • mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
Color Coded Caps
  • Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
  • Includes 5 color-coded replacement caps
Calibration Tool
  • Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
  • Can also be used as a tube opener
Specifications
Specifications
Color Coded Caps
mLINE™/Proline™ Plus mechanical pipette
Gray/Yellow/Blue/Green/Red
Color coding mLINE pipets by volume
5/Pk.
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021