Sartorius™ Biohit™ Pipet Stands and Accessories

Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius™ Biohit™ Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.

Manufacturer: Sartorius™ 725600

 View more versions of this product

Catalog No. 14-559-418

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Description Carousel Pipet Stand
    For Use With (Application) Storing pipets when not in use
    Quantity 1/Ea.
    Color White
    Number of Channels Holds 6 Pipets
    For Use With (Equipment) mLINE™/Proline™ Plus mechanical pipette
    Product Type Bioht™ carousel stand
    Holds 6 Pipetters
    No. of Positions 6.0

    Pipette Stands

    • Stores pipettes in an upright position while not in use to avoid contamination from the desktop
      • Linear Stand is designed for all Biohit mechanical devices and stores them neatly in a line for instant access
      • Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
      • mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
    Color Coded Caps
    • Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
    • Includes 5 color-coded replacement caps
    Calibration Tool
    • Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
    • Can also be used as a tube opener

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit