Sartorius™ Biohit PROLINE™ Plus Pipetter Starter Kits
Comfortable handle and finger support for minimized risk of strain injuries
Manufacturer: Sartorius™ 728652
 View more versions of this product
Catalog No. 14-559-457
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Finger support for minimized risk of strain injuries
    Includes Three single channel pipette (2 to 20μL, 20 to 200μL, 100 to 1000μL), optifit tip (0.5 to 200μL, 10 to 1000μL), single tray, three pipette holders, three calibration,Three single channel pipette (2 to 20μL, 20 to 200μL, 100 to 1000μL), optifit tip (0.5 to 200μL, 10 to 1000μL), single tray, three pipette holders, three calibration
    Product Type Proline™ Plus pipette starter kit 3
    Type Proline™ Plus pipette starter kit 3
  • Description & Specifications
Related Products
Product Content Feedback