Sartorius™ Biosart™ 100 Nutrient Media Available on GSA/VA Contract for Federal Government customers only.

Microbiological Nutrient Media for Filtration Units

Manufacturer: Sartorius™ 1640002WLK

 View more versions of this product

Catalog No. 14-555-775

  Check Availability
Add to Cart

    Specifications

    Detects Enterococci
    For Use With (Application) Microbiological testing of pharmaceuticals, food, beverages, Water and other liquids
    Format Liquid
    Manufacturing Origin United States
    Media Type WL Differential ; Wallerstein Differentia
    Packaging Ampule
    Quantity 2.5mL
    Shelf Life 1 Year
    Storage Requirements 4°C
    Certifications/Compliance DIN, ISO

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit