Sartorius™ BIOSTAT A™ Advanced Equipment Packages, for Cell Culture Applications
Entry-level bioreactor/fermenter ideal for educational purposes
Manufacturer: Sartorius™ CC1A
 View more versions of this product
Catalog No. 14-558-127
 N/A
N/A
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or email to cs.webemail@thermofisher.com to discuss your specific requirements. 
    Specifications
    Product Type Bioreactor package
    Capacity 1L
    For Use With (Application) Cell culture
    For Use With (Equipment) Biostat A
    Includes Advanced version for substrate controller/external feed pump; Operation via tablet and smartphone; 2 x spare analog inputs
  • Description & Specifications
Related Products
Product Content Feedback