Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Cellulose Acetate Membrane Filters

Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius™ Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.

Manufacturer:  Sartorius™ 1110790G

 View more versions of this product

Catalog No. 14-555-371


Add to cart

Description

Description

  • Ideal for colony counting, particle testing, microscopy and sterility testing
  • High-quality CA membrane filters for use in pressure filtration devices
  • Available in pore sizes ranging from 0.2µm up to 5.0µm
    • 0.2µm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
    • 5µm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
  • White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
  • Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide

The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.

Specifications

Specifications

0.2μm
Cellulose Acetate
White
Yes
120µm
Non-sterile
90mm
25/pk.
For use in pressure filtration devices; Sterile filtration of solution.
Hydrophillic
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit