Sartorius™ Combisart™ Individual Systems for Colony Counting Available on GSA/VA Contract for Federal Government customers only.
Sterile venting of every filter station
Manufacturer: Sartorius™ 16844
 View more versions of this product
Catalog No. 14-555-802
  Check Availability
Add to Cart
    Specifications
    Capacity 0mL
    Certifications/Compliance ISO 8199, B.S. 304S31, AISI 304
    Diameter (English) 1.85/1.96 in.
    Diameter (Metric) 4.7/5cm
    Filtration Area 12.5cm2
    Inlet Style Female Threads, TR 20 x 2
    Material Stainless Steel
    Outlet Style Hose Nipple, DN 10
    Sample Size 3 or 6
    For Use With (Application) Microbiological analysis or particle count in quality assurance
    Product Type 1 branch manifold without funnels and lids
  • Description & Specifications
Related Products
Product Content Feedback