Learn More
Sartorius™ Competence™ CPA Analytical Balances
Combines high-performance technological features and a rugged, built-to-last design
Manufacturer:  Sartorius™ CPA225D
 View more versions of this product
Catalog No. 14-557-036
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Analytical Balance
ISO/GLP
13.46 in.
0.01 / 0.1mg
9.13 in.
Manual Glass draft shield
Manual
±1ppm/°C
41 to 104°F
230/115V
7.6kg
230/115V 40/60Hz
2 Years
RS232C Bidirectional
34cm
CPA225D
8.3 in.
16 (maximum)/8 (typical)ppm/K
200g
34.2cm
LCD
40/60Hz
23.2cm
<6.3 sec.
≤6 / 3 sec.
5 to 40°C
16.75 lb.
8cm
50g
40 / 100 / 220g
13.38 in.
<±0.03, 0.1, 0.2mg
<±0.02, 0.05, 0.1mg
21.3cm