Learn More
Sartorius™ Complete Assembly for Vivacell 250
250 ml - 600 µL Vivacell 250 complete with pressure head, pressure indicator/over-pressure safety valve, quick-release connection to APC, 2 sample reservoirs, 2 filtrate containers and insert tool (no inserts included; sold separately).
Manufacturer:  Sartorius™ VCA250
Catalog No. 14-558-300
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Pressure head, pressure indicator/over-pressure release valve, quick release connection to APC, two sample reservoirs, filtrate container and insert tool
Description
Description
  • Easy to handle
  • High flux PES membrane
  • Vertical membrane design for ultrafast concentration
  • High retentate recovery, >90 %
  • Unit reusable, only membrane insert needs to be replaced
Specifications
Specifications
Polyethersulfone
40cm2
116mm
Pressure head; pressure indicator/over-pressure release valve; quick release connection to APC; two sample reservoirs; filtrate container; insert tool
250mL
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021