Learn More
Sartorius™ Cubis™ Essential Semimicro Balances
The right choice for users without complex operations who primarily want to perform ultra-precise weighing. Sartorius Cubis Essential Semimicro Balances have robust, monolithic weighing system technology for reliable measuring results.
Manufacturer:  Sartorius™ MSE225SED15
Catalog No. 14-559-000
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Features
  • Large, high-contrast, liquid crystal Cubis MSE display
  • Easy-to-understand menu guidance with short text prompts
  • Clearly structured keys for precise activation of functions
  • Software support for fast leveling
  • Easy calibration with isoCAL (internal automatic calibration | adjustment function)
  • Amply dimensioned, rectangular weighing pans
  • Data Interfaces: USB and RS232C; 3 optional ports are available as an accessory (Bluetooth, PS/2, RS232C)
  • Choose from 13 preconfigured premium Cubis Lab Balances, including semi-micro, analytical, precision and high-capacity balances
Draft Shields
  • Semi-micro and analytical models (14-559-000, 001, 002) include draft shields with high mechanical stability and effortlessly silent moving doors.
  • No disturbing frame, for unhindered access to the weighing pan.
  • Protected against electrostatic influences by a conductive coating
Applications
Unit conversion, isoCAL automatic calibration/adjustment function, density determination (buoyancy method only), calculations, averaging, net/total formulation, weighing in percent, counting,
totalizing.
Specifications
Specifications
85mm
Slotted weighing pan; separate electronics box
85mm
MSE225S-ED15
0.01 mg
≤ 6 sec.
≤ 0.1mg
0-60g: 0.015mg; 60-220g: 0.025mg
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021