Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cubis™ Ultra Micro Balances, 2.7S Weighing Mode with DM Draft Shield

Designed to weigh the smallest sample quantities and meet the highest requirements

Manufacturer:  Sartorius MSU2.7S000DM

 View more versions of this product

Catalog No. 14-557-805


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Specifications

2.1g
1A
Keys
16.1 in.
≤±0.0009mg
0.0001mg
≤±0.00025mg
14.6 in.
1.5cm
50/60Hz
Semi automatic
≤7 sec.
100/240V
9.8 in.
5cm
Gram, kilogram, carat, pound, ounce, troy ounce, Hong Kong tael, Singapore tael, Taiwan tael, grain, pennyweight, milligram
80% for 31°C, 50% for 40°C
41 to 104°F
Ultra micro balance
Internal, isoCAL
ISO, FDA/USP, GxP, USP31-NF26
41cm
RS232, USB, Ethernet, SD Card
MSU2.7S-000-DM
Color Graphic Display
0.59 in.
37.1cm
Automatic round Glass draft shield
≤10 sec.
1g
7w
24.9cm
100/240V 50/60Hz
5 to 40°C
1mg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit