Sartorius™ CultiBag RM Optical Available on GSA/VA Contract for Federal Government customers only.
Optical bags feature single use optochemical pH and DO sensor, which are already installed in everybag and calibrated
Manufacturer: Sartorius™ DBO050L
 View more versions of this product
Catalog No. 14-558-218
 N/A
    N/A
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or email to cs.webemail@thermofisher.com to discuss your specific requirements. 
    Specifications
    Product Type Cultibag
    Material EVA
    Model CultiBag RM 50L Optical
    Capacity 13.2 gal., 50L
    For Use With Used in seed train and production applications without ph and do control, biostat™ rm control tower
    Disposable Yes
    Ports Port A dip tube silicone, Port B C-Flex™ 374 tube, Port C septum, Port D DO sensor, Port E pH sensor, Port F silicone tube, Port G C-Flex™ 374 tube M-MPC, Port H outlet, Port I inlet
    Tubing Material C-Flex™ 374 Silicon
    Includes Air inlet/outlet filter, sample port, fill/drain and spare ports, female and male luer, female male mpc connectors, ports with dip tubes
    Capacity (English) 13.2 gal.
    Capacity (Metric) 50L
  • Description & Specifications
Related Products
Product Content Feedback