Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ Docu-pH+ Benchtop Meters

pH meter / mV meter

Manufacturer:  Sartorius™ DOCU-PHPLUS

 View more versions of this product

Catalog No. 14-557-210

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

Electrode retaining Arm, technical buffers, AC adapter, operating instructions
0.001/0.01/0.1
±2000mV
±0.02 <1000mV
Up to 500 data records
0 to 14pH
Automatic or Manual
0 to 14
±0.005
0.1/1mV
Color Graphic Display
pH
pH
-41° to +221°F
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit