Sartorius™ Docu-pH+ Benchtop Meters

pH meter / mV meter

Manufacturer: Sartorius™ DOCU-PHPLUS/P10DOC

 View all options for this product

Catalog No. 14-557-212


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Includes pH electrodes with Plastic body, refillable, fiber junction, NTC 10kw
    Range (pH) 0 to 14
    Resolution (pH) 0.001/0.01/0.1
    Accuracy (pH) ±0.005
    Range (mV) ±2000mV
    Resolution (mV) 0.1/1mV
    Accuracy (mV) ±0.2mV
    Display Type Color Graphic Display
    Data Management Details Up to 500 data records
    Outputs RS232
    Probe Type pH
    Weight (Metric) 0.06kg
    Depth (English) 9.1 in.
    Detection Range 0 to 14pH
    For Use With (Application) pH
    Height (English) 3.5 in.
    Height (Metric) 8.8cm
    Temperature Compensation Automatic or Manual
    Temperature Range (English) -41° to +221°F
    Weight (English) 2.2 lb.
    Width (Metric) 14.4cm
    Depth (Metric) 23.1cm

    DocuClip and Docu pH Meter let you close the reliability gap in electrochemistry. DocuClip and Docu pH Meter from Sartorius are setting new standards for documentation of results in electrochemistry.

    • Graphic display and soft keys for intuitive user guidance by plain-language prompts
    • Automatic temperature compensation, buffer recognition as well as recognition of various temperature sensors
    • Improved calibration routines support optimum procedural sequences in the laboratory
    • Calibration reminder
    • Memory for up to 500 data records
    • Data transfer over RS-232 to printer or PC
    • Automatic recognition of electrodes with DocuClip for unambiguous allocation of calibration data and complete documentation

    DocuClip & DocuMeter let you close the reliability gap in electrochemistry
    Laboratory work, particularly in the areas of chemical and pharmaceutical research, requires 100% traceable documentation. Conventional procedures, however, still hold some risks: Sensors that have been used may be mixed up. An electrode may have been used so often that it is no longer fully effective - but how can you tell? Manual recording of electrode identification is error-prone. Permanent allocation of identifying and calibration data to a particular pH electrode gives you complete reliability. It is no longer possible to mix up electrodes. The quality and effectiveness of electrodes can be checked at any time. Documentation of calibration data is fully automatic, making it both easy and reliable. It is entirely up to you whether you use your own electrodes or pre-calibrated electrodes from Sartorius. One click gives you complete reliability . . . with confidence supplied by Sartorius.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit