Learn More
Sartorius™ Electrode Refilling and storage solution 3M KCL
pH-buffer solutions and ready-to-use technical buffers for pH-calibration. The 250 and 1000 ml buffer kits of 4,7,10 color code assures the ability to singular dose necessary amount for daily pH-calibration routines. 50ml bottle of 3M KCI is required electrolyte refilling and the recommed storaging solution for all our pH electrodes.
Manufacturer:  Sartorius™ PYE502S
Catalog No. 14-557-905
Add to cart
Description
Description
  • A necessary accessory to maintain and keep liquid filled electrodes in optimal condition for accuracy and precision
  • Recommeded storage solution for all pH electrodes - keeps glass membranes hydrated and H+ ion activity fluid with high ionic concentrations
  • Reduces reference system exhuastion
Specifications
Specifications
pH Meters
50mL
Documents
Promotions
Promotions
  • Sartorius , Fisherbrand , Fisher Scientific
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021