Learn More
Sartorius™ Entris™ 90 mm Analytical Balances Available on GSA/VA Contract for Federal Government customers only.
Provides speedy results for heavy workloads with repetitive applications. Sartorius™ Entris™ 90mm Analytical Balances feature a new operating concept: easy configuration of individual settings guided by short, menu-driven text prompts and simple navigation using cursor keys.
Manufacturer:  Sartorius™ ENTRIS641S
 View more versions of this product
Catalog No. 14-557-448
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • Choice of 5 languages
  • External calibration only
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Description & Specifications
Specifications
90mm
External
2.5 sec.
330 x 230 x 330mm
0.2mg
0.1mg
Analytical Balance
0.1mg
176 x 173 x 240mm
60g
ENTRIS64-1S