Learn More
Sartorius Entris™ 90 mm Analytical Balances
Provides speedy results for heavy workloads with repetitive applications. Sartorius Entris™ 90mm Analytical Balances feature a new operating concept: easy configuration of individual settings guided by short, menu-driven text prompts and simple navigation using cursor keys.
Specifications
Readability 0.1 mg
Stabilization Time 2.5 sec.
View More Specs
Products 6
Catalog Number Mfr. No. Capacity Calibration Model Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Calibration Model Price Quantity & Availability  
14-557-447
View Documents
Sartorius
ENTRIS1241SUS
120 g No Internal Calibration ENTRIS124-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-557-446
View Documents
Sartorius
ENTRIS2241SUS
220 g External ENTRIS224-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-557-448
View Documents
Sartorius
ENTRIS641SUS
60 g External ENTRIS64-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-559-016
View Documents
Sartorius
ENTRIS64I1SUS
60 g Internal ENTRIS64i-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-559-018
View Documents
Sartorius
ENTRIS224I1SUS
220 g Internal ENTRIS224i-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
14-559-017
View Documents
Sartorius
ENTRIS124I1SUS
120 g Internal ENTRIS124i-1S This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • White-backlit, high-contrast display with 15mm digits; exceptionally easy to read under any room lighting conditions
  • Choice of 5 languages
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
0.1 mg
2.5 sec.