Learn More
Sartorius Funnel
$311.50
Specifications
Autoclavable Autoclavable
Diameter (Metric) Filter 4.7/5 cm
For Use With (Equipment) Combisart System
Type Funnel
For Use With (Application) Colony or particle counting
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
14-555-953
View Documents Promotion Details
Sartorius
6981065
100 mL
Each for $311.50
Add to cart
14-555-950
View Documents Promotion Details
Sartorius
6981004
40 mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
Specifications
Specifications
Autoclavable
Combisart System
Colony or particle counting
Stainless Steel
4.7/5 cm
Funnel
Closure clamp