Learn More
Sartorius™ Funnel Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 6981004
 View more versions of this product
Catalog No. 14-555-950
  Check Availability
Add to cart
Specifications
Description
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
Description & Specifications
Specifications
Yes
4.7/5cm
Closure clamp
Funnel
40mL
Colony or particle counting
Combisart™ system
Stainless Steel