Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ GoldLINE Electrodes

Superior stable results due to patented three chamber Iodine/Iodide metal-ion free reference system

Manufacturer:  Sartorius™ PYP292S/DEL

 View more versions of this product

Catalog No. 14-557-894

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Lowers temperature sensitivities and provides metal ion free reference system
  • Minimizes damage to sensors thereby elongating the life of the electrodes
  • Increases productivity and mitigates faulty measurements
  • Low resistance type A glass membrane
  • TRIS compatible and Impervious making it ideal for biological applications
Model PY-P28:
  • Pharma grade
  • pH/ATC
  • Glass body
  • Liquid filled 3M KCl
  • 3X Junction
  • Iodine/Iodide reference


Model PY-P29:
  • Micro (5mm dia.)
  • pH/ATC
  • Glass body
  • Liquid filled 3M KCl
  • 3X Junction
  • Iodine/Iodide reference


Model PY-P30:
  • Speartip
  • pH/ATC
  • Glass body
  • 3X Ceramic junction
  • Polymer filled
Applications:
Cosmetics, Agriculture, Dairy, Food and beverage, Surface (skin and textiles), Drinking water, Detergents agents, Pharmaceutical/Biotech/Clinical/Pharmacy, Biology/Microbiology and Chemistry Lab, Extreme and difficult samples
Specifications

Specifications

5mm
3X Junction
Glass
Iodine/Iodide
Documents
Promotions

Promotions

  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit