Learn More
Sartorius GoldLINE Electrodes
Superior stable results due to patented three chamber Iodine/Iodide metal-ion free reference system
Manufacturer: Sartorius PYP302S/DEL
Description
- Lowers temperature sensitivities and provides metal ion free reference system
- Minimizes damage to sensors thereby elongating the life of the electrodes
- Increases productivity and mitigates faulty measurements
- Low resistance type A glass membrane
- TRIS compatible and Impervious making it ideal for biological applications
Model PY-P28:
- Pharma grade
- pH/ATC
- Glass body
- Liquid filled 3M KCl
- 3X Junction
- Iodine/Iodide reference
Model PY-P29:
- Micro (5mm dia.)
- pH/ATC
- Glass body
- Liquid filled 3M KCl
- 3X Junction
- Iodine/Iodide reference
Model PY-P30:
- Speartip
- pH/ATC
- Glass body
- 3X Ceramic junction
- Polymer filled
Cosmetics, Agriculture, Dairy, Food and beverage, Surface (skin and textiles), Drinking water, Detergents agents, Pharmaceutical/Biotech/Clinical/Pharmacy, Biology/Microbiology and Chemistry Lab, Extreme and difficult samples
Specifications
Not Applicable | |
3X Hole Junction | |
Glass | |
Not Applicable |
Promotions
-
Sartorius
,
Fisherbrand
,
Fisher Scientific
Get a Pipette Multipack with Purchase
Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.: 06-30-2021