Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius GoldLINE Electrodes

Superior stable results due to patented three chamber Iodine/Iodide metal-ion free reference system

Specifications

Material Glass
View More Specs

Products 3
Catalog Number Mfr. No. dimType Junction Reference Type FillingSolutionType Price Quantity & Availability  
Catalog Number Mfr. No. dimType Junction Reference Type FillingSolutionType Price Quantity & Availability  
14-557-893
View Documents
Sartorius
PYP282S/DEL
3X Junction Iodine/Iodide This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-557-894
View Documents
Sartorius
PYP292S/DEL
3X Junction Iodine/Iodide This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-557-895
View Documents
Sartorius
PYP302S/DEL
3X Hole Junction Not Applicable This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Lowers temperature sensitivities and provides metal ion free reference system
  • Minimizes damage to sensors thereby elongating the life of the electrodes
  • Increases productivity and mitigates faulty measurements
  • Low resistance type A glass membrane
  • TRIS compatible and Impervious making it ideal for biological applications
Model PY-P28:
  • Pharma grade
  • pH/ATC
  • Glass body
  • Liquid filled 3M KCl
  • 3X Junction
  • Iodine/Iodide reference


Model PY-P29:
  • Micro (5mm dia.)
  • pH/ATC
  • Glass body
  • Liquid filled 3M KCl
  • 3X Junction
  • Iodine/Iodide reference


Model PY-P30:
  • Speartip
  • pH/ATC
  • Glass body
  • 3X Ceramic junction
  • Polymer filled
Applications:
Cosmetics, Agriculture, Dairy, Food and beverage, Surface (skin and textiles), Drinking water, Detergents agents, Pharmaceutical/Biotech/Clinical/Pharmacy, Biology/Microbiology and Chemistry Lab, Extreme and difficult samples
Specifications

Specifications

Glass
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit