Learn More
Sartorius™ Gridded Nonsterile Cellulose Nitrate Membrane Filters: 0.45µm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 1380647R
 View more versions of this product
Catalog No. 5090117774
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.45μm
47mm
No
For colony count; sterility testing; particle count; microscopy
Cellulose Nitrate
Green
1,000/pk.
Dark Green