Learn More
Sartorius™ Gridded Nonsterile Cellulose Nitrate Membrane Filters: 0.45µm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 1380650N
 View more versions of this product
Catalog No. 5090117776
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0.45μm
50mm
No
For colony count; sterility testing; particle count; microscopy
Cellulose Nitrate
Green
100/pk.
Dark Green