Learn More
Sartorius™ Gridded Nonsterile Cellulose Nitrate Membrane Filters: 1.2µm Pore Size Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sartorius™ 1140350N
 View more versions of this product
Catalog No. 5090117741
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1.2μm
50mm
No
For colony count; sterility testing; particle count; microscopy
Cellulose Nitrate
White
100/Pk.
Black