Learn More
Sartorius™ Low Retention SafetySpace™ Filter Tips
Maximize sample recovery, pipetting precision and sample security. Sartorius™ Low Retention SafetySpace™ Filter Tips feature a liquid-repellent surface, additive-free filter and a perfect-sealing fit. Compatible with Sartorius Biohit™ pipettes. Sterile tips in a range of volumes are packed in single trays.
Manufacturer:  Sartorius™ LHLF790011
 View more versions of this product
Catalog No. 14-557-693
  Check Availability
Add to cart
Specifications
Description
Low-retention tips for maximum sample recovery
  • Extremely even, liquid-repellent and hydrophobic surface optimizes the flow of liquids
  • Liquids slide easily off the chemically resistant surface, leaving very little residue behind
  • Ideal for fluids with low surface tension, such as viscous, detergent-containing liquids
  • Useful in PCR/real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods
Maximum safety and highest quality
  • Features a polyethylene filter without self-sealing additives that can interfere with the sample and results
  • Safety air gap between sample and filter reduces the risk of sample absorption into the filter
  • Filter protects against aerosol and liquid contamination and reduces pipette maintenance
  • E-beam pre-sterilized for sterile work
  • Certified DNase, RNase and endotoxin-free and compliant with ISO 9001 and ISO 13485
Ergonomically designed for precision and convenience
  • Perfect tip fitting and sealing to provide maximum precision and accuracy
  • Enables ergonomic and light tip attachment/ejection with Optiload feature of Sartorius pipettes
  • Compatible with Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipettes and most other brands
  • Color-coded trays and tips for easy matching with compatible Sartorius pipettes
  • Virgin polypropylene is 100% recyclable or can be incinerated as energy waste
Description & Specifications
Specifications
SafetySpace Filter Tips, Low Retention
Yes
Gray
Polypropylene tip, polyethylene filter
10 x 96
DNase, RNase and endotoxin free
E-beam
0.1 to 10μL
Sterile
Yes
Single Tray
3.2cm
Biohit™ pipettes and most other pipette brands