Sartorius™ Low Retention SafetySpace™ Filter Tips

Maximize sample recovery, pipetting precision and sample security. Sartorius™ Low Retention SafetySpace™ Filter Tips feature a liquid-repellent surface, additive-free filter and a perfect-sealing fit. Compatible with Sartorius Biohit™ pipettes. Sterile tips in a range of volumes are packed in single trays.

Manufacturer: Sartorius™ LHLF791001

 View more versions of this product

Catalog No. 14-557-698

  Check Availability
Add to Cart

    Specifications

    Sterility Sterile
    Product Type SafetySpace Filter Tips, Low Retention
    Material Polypropylene tip, polyethylene filter
    Quantity 10 x 96
    Length (Metric) 7.8cm
    For Use With (Equipment) Biohit™ pipettes and most other pipette brands
    Clean Claims DNase, RNase and endotoxin free
    Sterilization Method E-beam
    Filtered Yes
    Autoclavable Yes
    Packaging Single Tray
    Volume (Metric) 50 to 1000μL
    Color Code Blue

    Low-retention tips for maximum sample recovery

    • Extremely even, liquid-repellent and hydrophobic surface optimizes the flow of liquids
    • Liquids slide easily off the chemically resistant surface, leaving very little residue behind
    • Ideal for fluids with low surface tension, such as viscous, detergent-containing liquids
    • Useful in PCR/real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods
    Maximum safety and highest quality
    • Features a polyethylene filter without self-sealing additives that can interfere with the sample and results
    • Safety air gap between sample and filter reduces the risk of sample absorption into the filter
    • Filter protects against aerosol and liquid contamination and reduces pipette maintenance
    • E-beam pre-sterilized for sterile work
    • Certified DNase, RNase and endotoxin-free and compliant with ISO 9001 and ISO 13485
    Ergonomically designed for precision and convenience
    • Perfect tip fitting and sealing to provide maximum precision and accuracy
    • Enables ergonomic and light tip attachment/ejection with Optiload feature of Sartorius pipettes
    • Compatible with Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipettes and most other brands
    • Color-coded trays and tips for easy matching with compatible Sartorius pipettes
    • Virgin polypropylene is 100% recyclable or can be incinerated as energy waste

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit