Sartorius™ M-Power™ Precision Balances 150g to 6100g capacity

The clarity of the display and the one-key-to-a-function design of the keypad ensure error-free operation

Manufacturer: Sartorius™ AZ612

 View more versions of this product

Catalog No. 14-557-103

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Capacity (Metric) 610g
    Readability 0.01g
    Model AZ612
    Material Durable ABS housing
    Stabilization Time 2.5 sec.
    Type Precision Balance
    Connection Type RS232

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit