Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ M-Power™ Precision Balances 150g to 6100g capacity

The clarity of the display and the one-key-to-a-function design of the keypad ensure error-free operation

Manufacturer:  Sartorius™ AZ3102

 View more versions of this product

Catalog No. 14-557-105

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Specifications

Precision Balance
0.01g
Durable ABS housing
3100g
RS232
AZ3102
2.5 sec.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit