Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ MGC Grade Glass Microfiber Filters w/o Binder

Manufacturer:  Sartorius™ FT31103090

 View more versions of this product

Catalog No. 14-555-160


Add to cart

Description

Description

Applications include: Analysis of suspended solids in wastewater according to EN 872.
Specifications

Specifications

51mbar
3.5 in.
Filter Disc
1.2μm
932/1022°F
0.009 in.
9cm
Borosilicate Microfiber Glass
White
66 sec.
MGC
Non-sterile
500/550°C
0.25mm
Ideal for critical analyses in which impurities can be washed out of the filter
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit