Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ MGF Grade Glass Microfiber Filters w/o Binder

Manufacturer:  Sartorius™ FT31105142

 View more versions of this product

Catalog No. 14-555-135


Add to cart

Description

Description

Application Examples: Clarification of protein solutions, Toxicity Characteristic Leaching Procedure (TCLP) Test.
Specifications

Specifications

145mbar
5.59 in.
MGF
Non-sterile
500°C
0.4mm
Critical analyses in which impurities can be washed out of the filter
White
Filter Disc
0.7μm
932°F
0.015 in.
14.2cm
Borosilicate Microfiber Glass
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit