Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius™ MGG Grade Glass Microfiber Filters w/o Binder

Manufacturer:  Sartorius™ FT31106100

 View more versions of this product

Catalog No. 14-555-123

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ideal for critical analyses in which impurities can be washed out of the filter.
Specifications

Specifications

32mbar
White
Ideal for critical analyses in which impurities can be washed out of the filter
Filter Disc
1.5μm
0.01 in.
3.93 in.
Non-sterile
Borosilicate Microfiber Glass
10cm
38 sec.
MGG
500°C
0.28mm
932°F
Documents
Promotions

Promotions

  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.

    06-30-2021

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit